O-Methylhydroxylamine hydrochloride ∼40% ∼5.4 mol/L in water

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

M0886-25ML M0886-500ML
TCIAM0886-25MLEA 41.8 EUR
TCIAM0886-25ML TCIAM0886-500ML
O-Methylhydroxylamine hydrochloride ∼40% ∼5.4 mol/L in water
O-Methylhydroxylamine hydrochloride
Formule: CH₆ClNO
MDL Nummer: MFCD00012951
CAS nummer: 593-56-6
EINECS: 209-798-7
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cd506adfb72dcdf1523f1f9e591801ae08b719c&vid=0a70e6a58bf3a9d44b3655e2255baeb73e3e4314&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Concentration (Neutralization titration) 40.0 to 44.0 w/w%

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR