2-Methyl-1,3-cyclopentanedione ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M0901-25G M0901-5G
TCIAM0901-25GEA 106 EUR
TCIAM0901-25G TCIAM0901-5G
2-Methyl-1,3-cyclopentanedione ≥97.0% (by GC, titration analysis)
2-Methyl-1,3-cyclopentanedione
Formule: C₆H₈O₂
Kookpunt: 213 °C (1013 hPa)
Dichtheid: 1,33 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001406
CAS nummer: 765-69-5
EINECS: 212-153-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11bb2f3e0dddeac7bbdff10851747aef8c559530&vid=11bb2f3e0dddeac7bbdff10851747aef8c559530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 210.0 to 217.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR