2-Methoxybenzyl alcohol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

M1017-25G
TCIAM1017-25GEA 42.9 EUR
TCIAM1017-25G
2-Methoxybenzyl alcohol ≥98.0% (by GC)
2-Methoxybenzyl alcohol
Formule: C₈H₁₀O₂
Kookpunt: 250 °C (1013 hPa)
Dichtheid: 1,12 g/cm³ (20 °C)
Vlampunt: 128°C
MDL Nummer: MFCD00004611
CAS nummer: 612-16-8
EINECS: 210-296-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33795a4f2d7612c8c7c0e49fd45242e38521e3a1&vid=33795a4f2d7612c8c7c0e49fd45242e38521e3a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR