5-Nitro-2-pyridinethiol ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M1102-5G M1102-25G
TCIAM1102-5GEA 57.2 EUR
TCIAM1102-5G TCIAM1102-25G
5-Nitro-2-pyridinethiol ≥98.0% (by titrimetric analysis)
5-Nitro-2-pyridinethiol
Formule: C₅H₄N₂O₂S
MDL Nummer: MFCD00030890
CAS nummer: 2127-09-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d16da7111d9c789d630ceb80f87b09d24c0f227&vid=8d16da7111d9c789d630ceb80f87b09d24c0f227&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Methanol almost transparency

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR