Methyl-4,6-O-benzylidene-ɑ-D-glucopyranoside ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Methyl 4,6-O-Benzylidene-α-D-glucopyranoside, (4aR,6S,7R,8R,8aS)-6-Methoxy-2-phenylhexahydropyrano[3,2-d][1,3]dioxine-7,8-diol

M1125-25G M1125-5G
TCIAM1125-25GEA 217 EUR
TCIAM1125-25G TCIAM1125-5G
Methyl-4,6-O-benzylidene-ɑ-D-glucopyranoside ≥98.0% (by GC)
Methyl-4,6-O-benzylidene-alpha-D-glucopyranoside
Formule: C₁₄H₁₈O₆
MW: 282.29 g/mol
MDL Nummer: MFCD00006819
CAS nummer: 3162-96-7
EINECS: 221-615-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Specific rotation [a]20/D +100.0 to +110.0 deg (C=2, CH3CN)
Drying loss max. 4.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR