5-Methoxyindole-2-carboxylic acid ≥97.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M1271-25G M1271-5G
TCIAM1271-25GEA 240 EUR
TCIAM1271-25G TCIAM1271-5G
5-Methoxyindole-2-carboxylic acid ≥97.0% (by HPLC, titration analysis)
5-Methoxyindole-2-carboxylic acid
Formule: C₁₀H₉NO₃
MDL Nummer: MFCD00005614
CAS nummer: 4382-54-1
EINECS: 224-487-6

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR