(Methoxycarbonylmethyl)triphenylphosphonium bromide ≥97.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

M1326-25G
TCIAM1326-25GEA 82.5 EUR
TCIAM1326-25G
(Methoxycarbonylmethyl)triphenylphosphonium bromide ≥97.0% (by HPLC, titration analysis)
(Methoxycarbonylmethyl)triphenylphosphonium bromide
Formule: C₂₁H₂₀BrO₂P
MDL Nummer: MFCD00011801
CAS nummer: 1779-58-4
EINECS: 217-222-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0284cd746137a0b810d20d1147229cb208e1c77a&vid=0284cd746137a0b810d20d1147229cb208e1c77a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Precipitation Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR