2-(2-Methylphenyl)ethanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M1375-25G M1375-5G
TCIAM1375-25GEA 258 EUR
TCIAM1375-25G TCIAM1375-5G
2-(2-Methylphenyl)ethanol ≥98.0% (by GC)
2-(2-Methylphenyl)ethanol
Formule: C₉H₁₂O
Kookpunt: 243°C
MDL Nummer: MFCD00044241
CAS nummer: 19819-98-8
EINECS: 243-349-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=780ceca5371290bcbc21cc8d165a531e6ed57f86&vid=780ceca5371290bcbc21cc8d165a531e6ed57f86&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR