3-(Methylamino)-1-propanol ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

M1484-25G M1484-5G
TCIAM1484-25GEA 293 EUR
TCIAM1484-25G TCIAM1484-5G
3-(Methylamino)-1-propanol ≥97.0% (by GC, titration analysis)
3-(Methylamino)-1-propanol
Formule: C₄H₁₁NO
Kookpunt: 169°C
MDL Nummer: MFCD06797456
CAS nummer: 42055-15-2
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR