4'-(Methylthio)acetophenone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M1672-25G M1672-5G
TCIAM1672-25GEA 175 EUR
TCIAM1672-25G TCIAM1672-5G
4'-(Methylthio)acetophenone ≥98.0% (by GC)
4'-(Methylthio)acetophenone
Formule: C₉H₁₀OS
MDL Nummer: MFCD00039835
CAS nummer: 1778-09-2
EINECS: 217-213-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31b5ae8b2b0a243a3074ad9589e3fdb37ca5dd72&vid=31b5ae8b2b0a243a3074ad9589e3fdb37ca5dd72&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 84.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR