Methyl-2,4-difluorobenzoate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

M1879-25G M1879-5G
TCIAM1879-25GEA 99 EUR
TCIAM1879-25G TCIAM1879-5G
Methyl-2,4-difluorobenzoate ≥98.0% (by GC)
Methyl-2,4-difluorobenzoate
Formule: C₈H₆F₂O₂
Kookpunt: 194°C
Vlampunt: 82°C
MDL Nummer: MFCD03093799
CAS nummer: 106614-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac4fe93e034d7ef084d28a0951aeebc572cc038f&vid=ac4fe93e034d7ef084d28a0951aeebc572cc038f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR