2-Methyl-6-(trimethylsilyl)phenyl trifluoromethanesulphonate ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

M1883-1G M1883-5G
TCIAM1883-1GEA 201 EUR
TCIAM1883-1G TCIAM1883-5G
2-Methyl-6-(trimethylsilyl)phenyl trifluoromethanesulphonate ≥96.0% (by GC)
2-Methyl-6-(trimethylsilyl)phenyl trifluoromethanesulphonate
Formule: C₁₁H₁₅F₃O₃SSi
CAS nummer: 556812-44-3
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR