Methyl trans-4-(aminomethyl)cyclohexanecarboxylate hydrochloride ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M1988-25G M1988-5G
TCIAM1988-25GEA 499 EUR
TCIAM1988-25G TCIAM1988-5G
Methyl trans-4-(aminomethyl)cyclohexanecarboxylate hydrochloride ≥98.0% (by GC, titration analysis)
Methyl trans-4-(aminomethyl)cyclohexanecarboxylate hydrochloride
Formule: C₉H₁₇NO₂
MDL Nummer: MFCD00797465
CAS nummer: 29275-88-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR