Methyl-5-bromonicotinate ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

M2008-1G M2008-5G
TCIAM2008-1GEA 17.6 EUR
TCIAM2008-1G TCIAM2008-5G
Methyl-5-bromonicotinate ≥98.0% (by GC, titration analysis)
Methyl-5-bromonicotinate
Formule: C₇H₆BrNO₂
MDL Nummer: MFCD00173668
CAS nummer: 29681-44-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=605ae8ca4b425c71af10d9898db33f5fc1387b2f&vid=605ae8ca4b425c71af10d9898db33f5fc1387b2f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 97.0 to 101.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR