Methyl-2,5-bis(trifluoromethyl)-3,6-dioxaundecafluorononanoate (mixture of isomers) ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M2030-5G
TCIAM2030-5GEA 96.8 EUR
TCIAM2030-5G
Methyl-2,5-bis(trifluoromethyl)-3,6-dioxaundecafluorononanoate (mixture of isomers) ≥95.0% (by GC)
Methyl-2,5-bis(trifluoromethyl)-3,6-dioxaundecafluorononanoate (mixture of isomers)
Formule: C₁₀H₃F₁₇O₄
Kookpunt: 155°C
MDL Nummer: MFCD00054723
CAS nummer: 26131-32-8

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 % (total of isomer)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR