4-Methyloctanoic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 4-methylcaprylic acid, (R,S)-(+,-)-4-Methyloctanoic acid

M2048-5G
TCIAM2048-5GEA 50.6 EUR
TCIAM2048-5G
4-Methyloctanoic acid ≥98.0% (by titrimetric analysis)
4-Methyloctanoic acid
Formule: C₉H₁₈O₂
Kookpunt: 143°C
MDL Nummer: MFCD00051938
CAS nummer: 54947-74-9
EINECS: 259-404-2
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71c6f5a86dd2f8e7fbc47bd0449cbd2e473019a8&vid=71c6f5a86dd2f8e7fbc47bd0449cbd2e473019a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR