Methyl-3-methyl-2-nitrobenzoate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Methyl 3-methyl-2-nitrobenzoate

M2213-5G M2213-25G
TCIAM2213-5GEA 22 EUR
TCIAM2213-5G TCIAM2213-25G
Methyl-3-methyl-2-nitrobenzoate ≥98.0% (by GC)
Methyl-3-methyl-2-nitrobenzoate
Formule: C₉H₉NO₄
MW: 195.17 g/mol
MDL Nummer: MFCD01003947
CAS nummer: 5471-82-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1908fd08b0b4252f9ca57cbd8f580cf5b5c19645&vid=1908fd08b0b4252f9ca57cbd8f580cf5b5c19645&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR