Methylmagnesium bromide ∼30% ∼3 mol/L in MeTHF (2-methyltetrahydrofuran)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

M2237-250G
TCIAM2237-250GEA 165 EUR
TCIAM2237-250G
Methylmagnesium bromide ∼30% ∼3 mol/L in MeTHF (2-methyltetrahydrofuran)
Methylmagnesium bromide
Formule: CH₃BrMg
MDL Nummer: MFCD00000041
CAS nummer: 75-16-1
EINECS: 200-844-1
UN: 3399
ADR: 4.3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2f68fe628e17ea211aef8be479c098480c05357&vid=c2f68fe628e17ea211aef8be479c098480c05357&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Concentration (sec Butanol method) 30.0 to 34.5 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR