2-(4-Methyl-2-thioxo-2,3-dihydrothiazol-5-yl)acetic acid ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

M2481-5G M2481-25G
TCIAM2481-5GEA 41.8 EUR
TCIAM2481-5G TCIAM2481-25G
2-(4-Methyl-2-thioxo-2,3-dihydrothiazol-5-yl)acetic acid ≥98.0% (by GC, titration analysis)
2-(4-Methyl-2-thioxo-2,3-dihydrothiazol-5-yl)acetic acid
Formule: C₆H₇NO₂S₂
MDL Nummer: MFCD01750430
CAS nummer: 34272-64-5
EINECS: 251-905-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ca8fb962916e96d6f9d0c06f0a6ab986fda2d56&vid=4ca8fb962916e96d6f9d0c06f0a6ab986fda2d56&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR