Methyl azetidine-3-carboxylate hydrochloride ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M2521-1G M2521-5G
TCIAM2521-1GEA 34.1 EUR
TCIAM2521-1G TCIAM2521-5G
Methyl azetidine-3-carboxylate hydrochloride ≥98.0% (by titrimetric analysis)
Methyl azetidine-3-carboxylate hydrochloride
Formule: C₅H₁₀ClNO₂
MDL Nummer: MFCD01861758
CAS nummer: 100202-39-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0dbff142e68a5ee2e0fd180087eafe802311f6ca&vid=0dbff142e68a5ee2e0fd180087eafe802311f6ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Purity (Argentmetric Titration) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR