Methyl-2-fluoro-5-nitrobenzenecarboxylate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

M3115-5G M3115-25G
TCIAM3115-5GEA 64.9 EUR
TCIAM3115-5G TCIAM3115-25G
Methyl-2-fluoro-5-nitrobenzenecarboxylate ≥98.0% (by GC)
Methyl-2-fluoro-5-nitrobenzenecarboxylate
Formule: C₈H₆FNO₄
Kookpunt: 139°C
MDL Nummer: MFCD06203689
CAS nummer: 2965-22-2

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR