Methyl-3-(benzylamino)propanoate ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M3187-1G M3187-5G
TCIAM3187-1GEA 35.2 EUR
TCIAM3187-1G TCIAM3187-5G
Methyl-3-(benzylamino)propanoate ≥98.0% (by GC, titration analysis)
Methyl-3-(benzylamino)propanoate
Formule: C₁₁H₁₅NO₂
Kookpunt: 147°C
MDL Nummer: MFCD01475856
CAS nummer: 23574-01-8

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR