Benzyl nicotinate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

N0083-25G
TCIAN0083-25GEA 23.1 EUR
TCIAN0083-25G
Benzyl nicotinate ≥98.0% (by GC)
Benzyl nicotinate
Formule: C₁₃H₁₁NO₂
Kookpunt: 189°C
Smeltpunt: 24 °C
Dichtheid: 1,17 g/cm³ (20 °C)
MDL Nummer: MFCD00023584
CAS nummer: 94-44-0
EINECS: 202-332-3
Merck Index: 14,06526
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59fb701c7e5ff3fdd7b8e36abfc5ba20c92a5168&vid=59fb701c7e5ff3fdd7b8e36abfc5ba20c92a5168&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR