(S)-ɑ-Methyl-4-nitrobenzylamine hydrochloride ≥99.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

N0543-1G
TCIAN0543-1GEA 47.3 EUR
TCIAN0543-1G
(S)-ɑ-Methyl-4-nitrobenzylamine hydrochloride ≥99.0% (by titrimetric analysis)
(S)-alpha-Methyl-4-nitrobenzylamine hydrochloride
Formule: C₈H₁₀N₂O₂.HCl
MDL Nummer: MFCD00066312
CAS nummer: 132873-57-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Argentmetric Titration) min. 99.0 %
Specific rotation [a]20/D -6.7 to -7.7 deg (C=1, 0.05mol/L NaOH sol.)
Solubility in Water faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR