1-Naphthyl boronic acid (contains varying amounts of Anhydride)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

N0630-1G N0630-25G N0630-5G
TCIAN0630-1GEA 19.8 EUR
TCIAN0630-1G TCIAN0630-25G TCIAN0630-5G
1-Naphthyl boronic acid (contains varying amounts of Anhydride)
1-Naphthyl boronic acid
Formule: C₁₀H₉BO₂
Smeltpunt: 259 °C
MDL Nummer: MFCD00019722
CAS nummer: 13922-41-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bae4c25ee0bed755c653a0507bdc7b6ab891bbc&vid=8bae4c25ee0bed755c653a0507bdc7b6ab891bbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light red powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR