10-Nonadecanone ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

N0658-25G N0658-5G
TCIAN0658-25GEA 318 EUR
TCIAN0658-25G TCIAN0658-5G
10-Nonadecanone ≥97.0% (by GC)
10-Nonadecanone
Formule: C₁₉H₃₈O
Kookpunt: 156°C
Smeltpunt: 57 °C
Dichtheid: 0,806 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009584
CAS nummer: 504-57-4
EINECS: 207-994-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72d00eaff15c0cdfae5f86e28d1985ecd212926f&vid=1b7fc4b04453d4d9ebd7fb83fc3abbb492a31b9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 58.0 to 62.0 deg-C
Solubility in Toluene very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR