3-Nitropyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

N0771-5G
TCIAN0771-5GEA 92.4 EUR
TCIAN0771-5G
3-Nitropyridine ≥98.0% (by GC)
3-Nitropyridine
Formule: C₅H₄N₂O₂
Kookpunt: 216°C
Dichtheid: 1,33 g/cm³ (25 °C)
MDL Nummer: MFCD00234974
CAS nummer: 2530-26-9
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94e9f79929d602d5f873d29a14d9acf09d14521e&vid=94e9f79929d602d5f873d29a14d9acf09d14521e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR