4-Nitro-D-phenylalanine hydrate ≥97.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

N0849-1G N0849-5G
TCIAN0849-1GEA 27.5 EUR
TCIAN0849-1G TCIAN0849-5G
4-Nitro-D-phenylalanine hydrate ≥97.0% (by HPLC, titration analysis)
4-Nitro-D-phenylalanine hydrate
Formule: C₉H₁₀N₂O₄·H₂O
MDL Nummer: MFCD00150962
CAS nummer: 56613-61-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 % (calcd.on anh.substance)
Specific rotation [a]20/D -5.0 to -8.0 deg (C=2, 1mol/L HCl)
Water max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR