4-Nitro-L-phenylalanine methyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

N0878-1G N0878-5G
TCIAN0878-1GEA 17.6 EUR
TCIAN0878-1G TCIAN0878-5G
4-Nitro-L-phenylalanine methyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Nitro-L-phenylalanine methyl ester hydrochloride
Formule: C₁₀H₁₂N₂O₄
Smeltpunt: 224°C
MDL Nummer: MFCD00135754
CAS nummer: 17193-40-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +7.0 to +10.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR