5-Norbornene-2-carboxamide (mixture of isomers) ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

N0911-5G N0911-25G
TCIAN0911-5GEA 27.5 EUR
TCIAN0911-5G TCIAN0911-25G
5-Norbornene-2-carboxamide (mixture of isomers) ≥98.0% (by HPLC, total nitrogen)
5-Norbornene-2-carboxamide (mixture of isomers)
Formule: C₈H₁₁NO
Smeltpunt: 188°C
MDL Nummer: MFCD00187896
CAS nummer: 95-17-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 % (total of isomers)
Purity (with Total Nitrogen) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR