4-(2-Naphthyl)phenylboronic acid (contains varying amounts of Anhydride)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

N0946-200MG N0946-1G
TCIAN0946-200MGEA 22 EUR
TCIAN0946-200MG TCIAN0946-1G
4-(2-Naphthyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(2-Naphthyl)phenylboronic acid
Formule: C₁₆H₁₃BO₂
MDL Nummer: MFCD09260454
CAS nummer: 918655-03-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1950339f7d46fc286f8d5184afa694606bef234a&vid=1950339f7d46fc286f8d5184afa694606bef234a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 108.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR