Dimethylammonium dichlorotri(mu-chloro)bis{(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}diruthenate(II)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

N1024-200MG N1024-1G
TCIAN1024-200MGEA 39.6 EUR
TCIAN1024-200MG TCIAN1024-1G
Dimethylammonium dichlorotri(mu-chloro)bis{(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis{(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}diruthenate(II)
Formule: C₉₄H₉₆Cl₅NO₈P₄Ru₂
MDL Nummer: MFCD09753038
CAS nummer: 944451-14-3

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
IR to pass test

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR