Phenyl stearate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

O0007-25G O0007-5G
TCIAO0007-25GEA 448 EUR
TCIAO0007-25G TCIAO0007-5G
Phenyl stearate ≥98.0% (by GC)
Phenyl stearate
Formule: C₂₄H₄₀O₂
Kookpunt: 267°C
MDL Nummer: MFCD00026663
CAS nummer: 637-55-8
EINECS: 211-291-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62178a1556aa30faf37033a13b427ed0478e76c9&vid=62178a1556aa30faf37033a13b427ed0478e76c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR