Potassium oleate ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

O0056-25G O0056-500G
TCIAO0056-25GEA 17.6 EUR
TCIAO0056-25G TCIAO0056-500G
Potassium oleate ≥98.0% (by titrimetric analysis)
Potassium oleate
Formule: C₁₈H₃₃KO₂
Smeltpunt: 235…240 °C
MDL Nummer: MFCD00064243
CAS nummer: 143-18-0
EINECS: 205-590-5
Merck Index: 14,07650
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b58c153339ddf07faa27ba257662fdf963f08fc1&vid=b58c153339ddf07faa27ba257662fdf963f08fc1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR