2,2,3,3,4,4,5,5-Octafluoro-1-pentanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

O0114-25G O0114-500G O0114-100G
TCIAO0114-25GEA 17.6 EUR
TCIAO0114-25G TCIAO0114-500G TCIAO0114-100G
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol ≥98.0% (by GC)
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol
Formule: C₅H₄F₈O
Kookpunt: 140°C
Smeltpunt: < –50 °C
Dichtheid: 1,663 g/cm³ (20 °C)
MDL Nummer: MFCD00039631
CAS nummer: 355-80-6
EINECS: 206-593-4
Merck Index: 14,06746
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e05bb29f126986acd6fb29264be8b7bcab2d7366&vid=e05bb29f126986acd6fb29264be8b7bcab2d7366&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR