Octaethylene glycol monododecylether (C12E8) ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

O0139-5G O0139-1G
TCIAO0139-5GEA 626 EUR
TCIAO0139-5G TCIAO0139-1G
Octaethylene glycol monododecylether (C12E8) ≥97.0% (by GC)
Octaethylene glycol monododecylether (C12E8)
Formule: C₂₈H₅₈O₉
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00043234
CAS nummer: 3055-98-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=034df82bc195b9d1173c61226ed8da2f611f424b&vid=7e40f8230e2cd1f217154740340e6fe322481699&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR