Osmiumtetraoxide 4% in water

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

O0308-10ML
TCIAO0308-10MLEA 123 EUR
TCIAO0308-10ML
Osmiumtetraoxide 4% in water
Osmiumtetraoxide
Formule: O₄Os
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00011150
CAS nummer: 20816-12-0
EINECS: 244-058-7
UN: 3289
ADR: 6.1,II
Merck Index: 14,06893
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b39b04f7b5a3fdd69dd7e8d95a9e926d5d068f81&vid=767911f059a6d3a698d439fbff3bf3f4f0ad1251&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green clear liquid
Concentration 4.0 to 4.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR