Methyl palmitate ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Hexadecylic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Hexadecanoic acid methyl ester, Palmitic Acid Methyl Ester

P0006-25G P0006-250G
TCIAP0006-25GEA 17.6 EUR
TCIAP0006-25G TCIAP0006-250G
Methyl palmitate ≥97.0% (by GC)
Methyl palmitate
Formule: C₁₇H₃₄O₂
MW: 270.46 g/mol
Kookpunt: 417
Smeltpunt: 29.9
MDL Nummer: MFCD00008994
CAS nummer: 112-39-0
EINECS: 203-966-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38f70d63a93c1fb0816597b9de059bc2e020250a&vid=38f70d63a93c1fb0816597b9de059bc2e020250a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR