2-Pentene cis- and trans-mixture ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

P0064-25ML P0064-100ML
TCIAP0064-25MLEA 36.3 EUR
TCIAP0064-25ML TCIAP0064-100ML
2-Pentene cis- and trans-mixture ≥95.0% (by GC)
2-Pentene
Formule: C₅H₁₀
Kookpunt: 36°C
Smeltpunt: –140 °C
Dichtheid: 0,658 g/cm³ (20 °C)
Vlampunt: –45°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009384
CAS nummer: 109-68-2
EINECS: 203-695-0
UN: 3295
ADR: 3,II
Merck Index: 14,07123
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6fe84d62eb6b993ae0493e62fc01e1f4d6f7b43&vid=03c3a3497ce1a4793ad9e6c5f40b6d064de22159&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 % (total of isomers)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR