Phenol red 0,04% in water for pH determination

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

P0101-500ML
TCIAP0101-500MLEA 40.7 EUR
TCIAP0101-500ML
Phenol red 0,04% in water for pH determination
Phenol red
Formule: C₁₉H₁₄O₅S
MW: 354.38 g/mol
MDL Nummer: MFCD00003552
CAS nummer: 143-74-8
EINECS: 205-609-7
Merck Index: 14,07247
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e35941e5300ce3f9f380cfdacc7d6a498ebcb6a4&vid=eea2d04d7d67b0f9062b9106b1264156f8d89f23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light red to Yellow to Red clear liquid
Transition interval (pH) (yellow)6.8 to 8.4 (red)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR