2-Hydroxyiminopropiophenone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P0211-25G
TCIAP0211-25GEA 93.5 EUR
TCIAP0211-25G
2-Hydroxyiminopropiophenone ≥98.0% (by GC)
2-Hydroxyiminopropiophenone
Formule: C₉H₉NO₂
MDL Nummer: MFCD00002115
CAS nummer: 119-51-7
EINECS: 204-329-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4526bc4ae3fce9250c58bfdb037c430f04e9fcfb&vid=4526bc4ae3fce9250c58bfdb037c430f04e9fcfb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 112.0 to 115.0 deg-C
Solubility in Methanol almost transparency

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR