(±)-1-Phenyl-2-propyn-1-ol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 1-Phenyl-2-propyn-1-ol

P0220-5G
TCIAP0220-5GEA 69.3 EUR
TCIAP0220-5G
(±)-1-Phenyl-2-propyn-1-ol ≥98.0% (by GC)
(±)-1-Phenyl-2-propyn-1-ol
Formule: C₉H₈O
MW: 132.16 g/mol
Kookpunt: 115°C
MDL Nummer: MFCD00021860
CAS nummer: 4187-87-5
EINECS: 224-064-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc4b118285b0c5cd0fc098600ec4efecf0dcff5e&vid=bc4b118285b0c5cd0fc098600ec4efecf0dcff5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR