N,N,N-Trimethylanilinium bromide ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Phenyltrimethylammonium bromide, Trimethylphenylammonium bromide

P0243-25G P0243-500G P0243-100G
TCIAP0243-25GEA 22 EUR
TCIAP0243-25G TCIAP0243-500G TCIAP0243-100G
N,N,N-Trimethylanilinium bromide ≥98.0% (by HPLC, titration analysis)
N,N,N-Trimethylanilinium bromide
Formule: C₉H₁₄BrN
Smeltpunt: 214 °C
MDL Nummer: MFCD00011788
CAS nummer: 16056-11-4
EINECS: 240-202-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45d0e3ecea0f8007ef0b453ac823b59ec0adbaf3&vid=45d0e3ecea0f8007ef0b453ac823b59ec0adbaf3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR