Diisodecyl phthalate (mixture of branched chain isomers)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

P0299-25ML P0299-500ML
TCIAP0299-25MLEA 17.6 EUR
TCIAP0299-25ML TCIAP0299-500ML
Diisodecyl phthalate (mixture of branched chain isomers)
Diisodecyl phthalate
Formule: C₂₈H₄₆O₄
Vlampunt: 235°C
MDL Nummer: MFCD00048389
CAS nummer: 26761-40-0
EINECS: 247-977-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=070004447c8dbd4817bd780ff3dd97269e1d06ad&vid=070004447c8dbd4817bd780ff3dd97269e1d06ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Ester Value 246 to 254


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR