Potassium hydrogen phthalate ≥99.8% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Potassium phthalate monobasic, Potassium biphthalate, Potassium Acid Phthalate, KHP, Phthalic Acid Monopotassium Salt, Phthalic acid potassium salt

P0309-25G P0309-500G
TCIAP0309-25GEA 17.6 EUR
TCIAP0309-25G TCIAP0309-500G
Potassium hydrogen phthalate ≥99.8% (by titrimetric analysis)
Potassium hydrogen phthalate
Formule: HOOCC₆H₄COOK
MW: 204.22 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013070
CAS nummer: 877-24-7
EINECS: 212-889-4
Merck Index: 14,07612
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33dc082c9c7caaa1309254cdff228673bfd03d23&vid=33dc082c9c7caaa1309254cdff228673bfd03d23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.5 area%
Purity (Neutralization titration) 99.8 to 100.2 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR