Phenylthiohydantoin-δ-threonine ≥90.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P0370-1G P0370-100MG
TCIAP0370-1GEA 119 EUR
TCIAP0370-1G TCIAP0370-100MG
Phenylthiohydantoin-δ-threonine ≥90.0% (by HPLC)
Phenylthiohydantoin-delta-threonine
Formule: C₁₁H₁₀N₂OS
MDL Nummer: MFCD00070617
CAS nummer: 5800-50-0

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 90.0 area%
Melting point 234.0 to 238.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR