3-Picoline ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

P0416-25ML P0416-500ML
TCIAP0416-25MLEA 39.6 EUR
TCIAP0416-25ML TCIAP0416-500ML
3-Picoline ≥98.0% (by GC)
3-Picoline
Formule: C₆H₇N
Kookpunt: 144°C
Smeltpunt: –18°C
Dichtheid: 0,957 g/cm³ (20 °C)
Vlampunt: 36°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006402
CAS nummer: 108-99-6
EINECS: 203-636-9
UN: 2313
ADR: 3,III
Merck Index: 14,07401
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9f2470e5f677f021a46efa1deb820f37d2aba81&vid=e9f2470e5f677f021a46efa1deb820f37d2aba81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR