DL-Proline ≥99.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P0480-1G P0480-25G P0480-5G
TCIAP0480-1GEA 17.6 EUR
TCIAP0480-1G TCIAP0480-25G TCIAP0480-5G
DL-Proline ≥99.0% (by titrimetric analysis)
DL-Proline
Formule: C₅H₉NO₂
Kookpunt: 245 °C (1013 hPa)
Smeltpunt: 208…210 °C
Dichtheid: 1,38 g/cm³ (22.9 °C)
MDL Nummer: MFCD00005250
CAS nummer: 609-36-9
EINECS: 210-189-3
Merck Index: 14,07780
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8325448b726b813f96a1e8e734dab4cd303b1298&vid=8325448b726b813f96a1e8e734dab4cd303b1298&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR