Propyleencarbonaat ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

P0525-25G P0525-500G
TCIAP0525-25GEA 18.7 EUR
TCIAP0525-25G TCIAP0525-500G
Propyleencarbonaat ≥98.0% (by GC)
Propyleencarbonaat
Formule: C₄H₆O₃
Kookpunt: 242°C
Smeltpunt: -49°C
Dichtheid: 1,2 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005385
CAS nummer: 108-32-7
EINECS: 203-572-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd4f74a96ae6d9c8ed1a11d6e5993155eefbc650&vid=dd4f74a96ae6d9c8ed1a11d6e5993155eefbc650&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR