Ammonium 1-pyrrolidinecarbodithioate ≥95.0% (by titrimetric analysis) for atomic absorption spectroscopy

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Pyrrolidinedithiocarbamate ammonium salt, 1-Pyrrolidinecarbodithioic acid, ammonium salt, Ammonium pyrrolidine-1-carbodithioate, Pyrrolidine-1-Dithiocarboxylic Acid Ammonium Salt, APCD

P0644-25G P0644-250G
TCIAP0644-25GEA 40.7 EUR
TCIAP0644-25G TCIAP0644-250G
Ammonium 1-pyrrolidinecarbodithioate ≥95.0% (by titrimetric analysis) for atomic absorption spectroscopy
Ammonium 1-pyrrolidinecarbodithioate
Formule: C₅H₁₂N₂S₂
MW: 146.3 · 18.0 g/mol
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00012720
CAS nummer: 5108-96-3
EINECS: 225-834-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=678afd344c7ca5ee14b0f353166ff08d1bbf63ac&vid=b44558dcceffb3e162605437544ec2a3bd00d201&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 95.0 %
Solubility in Water very faint turbidity
Cadmium max. 1 ppm
Copper max. 1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR